Learn More
Medchemexpress LLC Bromo-PEG5-alcohol | 957205-14-0 | 98.0% | C10H21BrO5 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY141214500MG
Encompass_Preferred
Bromo-PEG5-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. PROTACs selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-53081