Learn More
Medchemexpress LLC Bromo-PEG6-alcohol | 136399-05-8 | ≥97.0% | C12H25BrO6 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1412151G
Encompass_Preferred
Bromo-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55816