Learn More
Medchemexpress LLC Bromo-PEG6-alcohol | 136399-05-8 | ≥97.0% | C12H25BrO6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141215100MG
Encompass_Preferred
Bromo-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • It is a PEG-based PROTAC linker.
  • It can be used in the synthesis of PROTACs.
  • PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
Catalog No. 50-003-55680