Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromo-PEG6-alcohol | 136399-05-8 | ≥97.0% | C12H25BrO6 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY141215

Encompass_Preferred

Bromo-PEG6-alcohol is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It is intended for research use only and not for patient use.

  • Functions as a polyethylene glycol (PEG)-based linker for the synthesis of Proteolysis Targeting Chimeras (PROTACs).
  • Specifically designed and sold for research applications.
  • Molecular Weight: 345.23.
  • Appearance: Liquid (Density: 1.282 g/cm3).
  • Color: Colorless to light yellow.

Catalog No. 50-003-52745


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.