Learn More
Medchemexpress LLC Bromo-PEG6-azide | 2062663-64-1 | 95.0% | C14H28BrN3O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14082350MG
Encompass_Preferred
Bromo-PEG6-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups and can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • For research use only
Catalog No. 50-003-55725