Learn More
Broadpharm Bromoacetamido-PEG2-azide, 500mg
Supplier:  Broadpharm BP22868500MG
Encompass
Bromoacetamido-PEG2-azide is a heterobifunctional PEG linker containing a reactive bromoacetamido and a terminal azide.The bromoacetamidogroup is a very reactive group for nucleophilic substitution. The azide group can react with alkyne, BCN, DBCO via Click Chemistry. Please contact us for GMP-grade inquiries.
Catalog No. NC3963574