Learn More
Medchemexpress LLC Bromoacetamido-PEG2-C2-acid | 1415800-44-0 | 99.8% | C9H16BrNO5 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141380
Encompass_Preferred
Bromoacetamido-PEG2-C2-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds employ two different ligands connected by a linker to harness the intracellular ubiquitin-proteasome system, enabling the selective degradation of specific target proteins. This product is intended for research use only.
  • PEG-based linker for PROTAC synthesis
  • Designed to connect two distinct ligands
  • Facilitates targeting of E3 ubiquitin ligase
  • Enables selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-55916