Learn More
Broadpharm Bromoacetamido-PEG23-azide, 1g
Supplier:  Broadpharm BP416941G
Encompass
Bromoacetamido-PEG23-azide ester is a bifunctional PEG linker with terminal bromoacetamido and azide groups. The bromoacetamido group partakes in nucleophilic substitution, while the azide group undergoes copper-catalyzed click reactions with alkyne, DBCO, BCN. The long PEG chain imparts hydrophilicity. Please contact us for GMP-grade inquiries.
Catalog No. NC3963564