Learn More
Medchemexpress LLC Bromoacetamido-PEG3-NH-Boc | 1421933-39-2 | C15H29BrN2O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1412005G
Encompass_Preferred
Bromoacetamido-PEG3-NH-Boc is a PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system.
  • Selectively degrade target proteins.
Catalog No. 50-003-53083