Learn More
Medchemexpress LLC Bromobenzene D5 | 4165-57-5 | MFCD00000056 | 5g
SDP
Supplier:  Medchemexpress LLC HYZ0056S5G
Encompass_Preferred
Bromobenzene-d5 is the deuterium labeled Bromobenzene Bromobenzene is a well-known environmental toxin which causes liver and kidney damage through CYP450-mediated bio-activation to generate reactive metabolites and consequently oxidative stress[1]
Catalog No. 50-002-66463