Learn More
Cayman Chemical BromocrIptIn-13C-d3 500ug
SDP
Supplier:  Cayman Chemical 26669500 UG
Encompass_Preferred
An internal standard for the quantification of bromocriptine by GC- or LC-MS
Catalog No. 50-293-6924