Learn More
Cayman Chemical Bromoenol lacton 10mg
SDP
Supplier:  Cayman Chemical 7070010 MG
Encompass_Preferred
BEL is a selective, irreversible, mechanism-based inhibitor of iPLA2 with a Ki value of 180 nM.{1631} It inhibits macrophage iPLA2 in a concentration-dependent manner with an IC50 value of 60 nM.{1291,1770}
Catalog No. 50-289-6515