Learn More
Medchemexpress LLC Bromoenol lactone 25mg | 88070-98-8 | 25MG
SDP
Supplier:  Medchemexpress LLC HY10741125MG
Encompass_Preferred
Bromoenol lactone ((6E)-Bromoenol lactone) is a suicide-based irreversible selective potent inhibitor of calcium-independent phospholipase A2 (iPLA2 ) with an IC50 value of approximately 7 M which inhibits antigen-stimulated mast cell exocytosis without blocking Ca2 influx[1 [2
Catalog No. 50-002-41355