Learn More
Strem, An Ascensus Company CAS# 344-04-7. 50g. Bromopentafluorobenzene, 99%. MFCD00000287
SDP
Supplier:  Strem, An Ascensus Company 09152050G
Encompass
CAS# 344-04-7. 50g. Bromopentafluorobenzene, 99%. MFCD00000287. Molecular Weight: 246.97. Molecular Formula: C6F5Br. Color/form: colorless liq. Strem# 09-1520. http://www.strem.com/catalog/v/09-1520/
Catalog No. 50-901-09281