Learn More
Medchemexpress LLC Bromophenol blue | 115-39-9 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB157110MM/1ML
Encompass_Preferred
Bromophenol blue is an acid phthalein dye primarily used as a tracking dye for electrophoresis and as a pH indicator with a transition range of pH 3 to 4.6. It also functions as a sensor for detecting substances like ammonia, drugs, proteins, and amino acids, showing a strong absorbance at 610 nm when bound to proteins. This product is intended for research use only.
  • Used as a tracking dye for electrophoresis.
  • Functions as a pH indicator with a transition range of pH 3 to 4.6.
  • Employed as a sensor for detecting substances such as ammonia, drugs, proteins, and amino acids.
  • Binds to proteins, leading to strong absorbance at 610 nm.
  • Can be prepared as a 0.0075% staining solution for use in protein assays.
Catalog No. 50-004-11726