Learn More
Medchemexpress LLC Bromophenol blue ind 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW110798
Encompass_Preferred
Bromophenol blue indicator (3 0-4 6) is a synthetic dye commonly used as an acid-base indicator with a transition range of pH 3 0-4 6 Bromophenol blue indicator (3 0-4 6) is water soluble and changes color from yellow to blue as the pH of the solution changes from acidic to basic Its unique chemical properties make it an important ingredient in a variety of scientific applications especially in biochemistry and molecular biology In addition it can be used as a stain in microbiology and histology However Bromophenol blue indicator (3 0-4 6) has potential irritating and staining properties
Catalog No. 50-002-05454