Learn More
Medchemexpress LLC Bromosalicylchloranilide | 3679-64-9 | 99.2% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB2140
Encompass_Preferred
Multifungin (Bromochlorosalicylanilide) is an antifungal agent used to treat oral candidiasis. It also prevents the formation and accumulation of Zearalenone and reduces the fungal population in stored-crushed corn. This product is for research use only and not sold to patients.
  • Interacts with bacterial cell membranes, changing intracellular H+ and K+ ion levels.
  • Destroys bacterial proton motive force (PMF), affecting respiratory chain function and ATP synthesis.
  • Inhibits phosphatidic acid biosynthesis, exerting an antibacterial effect.
  • Exhibits broad-spectrum antibacterial activity against various bacteria with MICs of 0.25-1 μg/mL.
  • Shows anti-infectious activity in infected mouse models, reducing bacterial load.
  • Exhibits anti-inflammatory activity in mouse peritonitis models, alleviating lung pathological changes.
Catalog No. 50-004-11637