Learn More
Selleck Chemical LLC Bromotetramisole Oxalate
SDP
Supplier:  Selleck Chemical LLC S738350MG
Encompass_Preferred
(-)-p-Bromotetramisole Oxalate (L-p-Bromotetramisole Oxalate) is a potent and non-specific alkaline phosphatase inhibitor
Catalog No. 50-000-01299