Learn More
Abcam Bromothymol blue, pH indicator, 25G
Supplier:  Abcam AB14629325G
Encompass_Preferred
MW 624.38 g/mol. pH indicator for weak acids and bases (pH range from 6.0 (yellow) to 7.6 (blue)). Also used as a stain to define cell walls or nuclei under the microscope.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4754