Learn More
Medchemexpress LLC Bromperidol hydrochloride | 59453-24-6 | 100.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0901A5MG
Encompass_Preferred
Bromperidol hydrochloride is an antipsychotic agent that has a high affinity for central dopamine receptors D2. It can also synergistically kill Mycobacteria when combined with Spectinomycin. For research use only.
  • Possesses antipsychotic activity due to its high affinity for central dopamine D2 receptors.
  • Can kill Mycobacteria synergistically with Spectinomycin.
  • Antagonizes stereotyped behavior and agitation induced by apomorphine or amphetamine in rats, and inhibits conditioned reactions and learned intracranial self-stimulation.
  • Antagonizes apomorphine-induced emesis and inhibits the conditioned avoidance response in dogs.
Catalog No. 50-004-08640