Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromperidol hydrochloride | 59453-24-6 | MFCD34676672 | 100.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0901A10MG

Encompass_Preferred

Bromperidol hydrochloride is the hydrochloride salt of bromperidol, an antipsychotic compound with high affinity for dopamine D2 receptors. Supplied as a solid research reagent for laboratory use, it is intended for in vitro studies and is not for human or veterinary use.

  • Hydrochloride salt of bromperidol.
  • Antipsychotic agent with high affinity for dopamine D2 receptors.
  • High purity: 99.96%.
  • Molecular formula: C21H24BrClFNO2.
  • Molecular weight: 456.78.
  • Appearance: solid.
  • For research use only; not for human or veterinary use.
  • Packaged as a 10 MG unit.

Catalog No. 50-002-98238


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.