Learn More
Apexbio Technology LLC Brompheniramine hydrogen maleate 980-71-2 50mg
SDP
Supplier:  Apexbio Technology LLC B154550
Encompass_Preferred
Brompheniramine hydrogen maleate (CAS 980-71-2) is a small molecule that acts as an antagonist of histamine H1 receptors By competitively inhibiting histamine binding at H1 sites it reduces the downstream effects of histaminergic signaling such as increased vascular permeability and smooth muscle contraction This compound is commonly utilized in research to study histamine-mediated physiological and pathological processes including models of allergy inflammation and related immune responses Its use enables investigation of the functional roles of H1 receptor blockade in various biological systems
Catalog No. 50-000-89999