Learn More
Cayman Chemical BromphenIramInmaleate 10mg
SDP
Supplier:  Cayman Chemical 2099610 MG
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 6.06 nM); also inhibits the reuptake of serotonin and norepinephrine
Catalog No. 50-298-1872