Learn More
Medchemexpress LLC Brophenexin free base | 1353979-43-7 | 99.2% | 257.18 g/mol | C11H17BrN2 | 5 MG

Supplier: Medchemexpress LLC HY139192A5MG
Brophenexin free base is a research small molecule that inhibits the NMDAR/TRPM4 interaction interface and has demonstrated neuroprotective activity in cellular and animal models, including prevention of NMDA-induced neuronal death (IC50 2.1 μM) and reduction of ischemic brain damage.
- Potent NMDAR/TRPM4 interaction interface inhibitor (IC50 2.1 μM).
- Demonstrated neuroprotective activity in vitro and in vivo.
- High purity (≈99.2%).
- Molecular formula C11H17BrN2; molecular weight 257.18 g/mol.
- Supplied as 5 mg for research use.
- Storage: pure form -20°C for long term; 4°C for shorter term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.