Learn More
Medchemexpress LLC Brophenexin free base | 1353979-43-7 | 99.2% | C11H17BrN2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY139192A10MG
Encompass_Preferred
Brophenexin free base is a research-grade small molecule inhibitor of the NMDAR-TRPM4 interaction interface with reported neuroprotective activity in cellular and animal models. It prevents NMDA-induced neuronal death, mitigates mitochondrial dysfunction, and modulates ERK1/2 and CREB-related signaling pathways.
  • High purity: 99.2% (rounded from 99.24%).
  • Molecular formula C11H17BrN2; molecular weight 257.18.
  • Demonstrated neuroprotective activity with reported IC50 of 2.1 μM.
  • Reduces GluN2A and GluN2B interactions with TRPM4, restoring ERK1/2 activation.
  • Available in small milligram quantities suitable for in vitro and in vivo studies.
  • Provided as the free base form for research applications.
Catalog No. 50-002-60966