Learn More
Perkin Elmer US LLC Brownlee Aquapore Prep-10 Octyl Cartridge Column - 250 mm x 4.6 mm I.D.
Supplier:  Perkin Elmer US LLC 07110166
Encompass_Preferred
The Aquapore Prep-10 Octyl cartridge column is based on a wide-pore, silica-based support suitable for the separation of large biopolymers. Each Brownlee Prep-10 cartridge incorporates a moveable inlet plug and filter, which compensates for changes in bed volume with continued use. The typical capacity of the 250 mm cartridge is 50 mg to 1 g, depending on the resolution and purity required. The typical flow rate is 4-10 mL/min. This cartridge column requires a holder: 250 mm Prep-10 Cartridge Holder (07150006) End-Capping: Yes Carbon Load: 5% Interaction Mode: Reversed Phase Surface Area: 100 m2/g
Catalog No. 50-211-3591