Learn More
Perkin Elmer US LLC Brownlee Pecosphere Aquapore Octyl C18 Cartridge Column - 83 mm x 4.6 mm I.D., 5 µm
Supplier:  Perkin Elmer US LLC 02580166
Encompass_Preferred
Monofunctional, non-reduced octyl (C18) phase recommended for the fast, reversed-phase separation of smaller compounds.
Catalog No. 50-211-3447