Learn More
Perkin Elmer US LLC Brownlee Pecosphere Spheri-5 C18 Scavenger Column - 33 mm x 4.6 mm I.D., Pkg. 5
Supplier:  Perkin Elmer US LLC 02580202
Encompass_Preferred
Brownlee Pecosphere Spheri-5 C18 Scavenger columns are packed with rugged, high purity silica with standard end-capping. The 10 µm particle size is designed to remove contaminants from the mobile phase when installed prior to the HPLC injector. These cartridge-style columns require the 33 mm fast LC Pecosphere cartridge holder (07150028). Quantity: Package of 5 End-Capping: Yes Carbon Load: 11% Interaction Mode: Reversed Phase pH Range: 2.0-8.0 Surface Area: 170 m2/g USP Code: L1
Catalog No. 50-211-3461