Learn More
Perkin Elmer US LLC Brownlee Spheri-10 RP-8 Cartridge Column - 100 mm x 4.6 mm I.D., Pkg. 2
Supplier:  Perkin Elmer US LLC 07110123
Encompass_Preferred
The Brownlee Spheri-10 RP-8 cartridge column has a small-pore, silica-based sorbent, optimized for separating small molecules. Also available in a 5 µm particle size (Spheri-5). This cartridge column requires a holder: To use with a guard - 100 mm NewGuard System Holder (07150016) To use without without a guard - 100 mm MPLC Holder (07150014) Quantity: Package of 2 End-Capping: Yes Carbon Load: 6% Interaction Mode: Reversed Phase pH Range: 2.5-8.0 Temperature Limit: 60 USP Code: L7
Catalog No. 50-211-3590