Learn More
Perkin Elmer US LLC Brownlee Spheri-5 RP-8 Cartridge Column - 220 mm x 4.6 mm I. D.
Supplier:  Perkin Elmer US LLC 07110005
Encompass_Preferred
Monofunctional bonded C8 phase for general purpose reversed phase small molecule applications.
Catalog No. 50-211-3544