Learn More
Perkin Elmer US LLC Brownlee SPP C18 Column - 150 mm x 2.1 mm I.D.
Supplier:  Perkin Elmer US LLC N9308405
Encompass_Preferred
High purity, general purpose Octadecyl (C18) phase with wide selectivity. Recommended for the reverse phase separation of acidic, basic, or neutral compounds.
Catalog No. 50-210-3932