Learn More
Perkin Elmer US LLC Brownlee SPP PhenylHexyl Column - 100 mm x 2.1 mm I.D.
Supplier:  Perkin Elmer US LLC N9308485
Encompass_Preferred
Alternative selectivity to alkyl bonded phases, recommended for aromatic groups. Compatible with highly aqueous mobile phases to facilitate the retention and separation of polar compounds.
Catalog No. 50-210-4032