Learn More
TARGETMOL CHEMICALS INC BRUCEANTINOL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN14535MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bruceantinol has antiviral activity it can inhibit pepper mottle virus in pepper; it shows antibabesial activity against Babesia gibsoni in vitro with the IC50 value of 12 ng/mL. purity: 99%
Catalog No. 50-257-8893