Learn More
TARGETMOL CHEMICALS INC BRUCEINE A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2S20465MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bruceine A (NSC-31616)(NSC-310616; Dihydrobrusatol) is a natural quassinoid compound extracted from the dried fruits of Brucea javanica (L.); are potential candidates for the treatment of canine babesiosis. purity: 99%
Catalog No. 50-257-7020