Learn More
Medchemexpress LLC Bruceine B | 25514-29-8 | MFCD34469149 | 99.6% | 480.46 | C23H28O11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN301310MG
Encompass_Preferred
Bruceine B is a bioactive natural product provided as a white to off-white solid for research use. It inhibits protein synthesis and nucleic acid synthesis and has reported antimalarial and cytotoxic activity in cellular assays.
  • Inhibits protein synthesis and nucleic acid synthesis.
  • High purity (99.6%) suitable for research applications.
  • Available in small milligram pack sizes for assay development and cell studies.
  • Includes supporting documents such as COA, SDS, HNMR, and LCMS.
  • Store protected from light at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-89430