Learn More
Bruker Daltonics BRUKER BACTERIAL TEST
Supplier: Bruker Daltonics 8255343
The BTS is an E.coli extract spiked with two high molecular weight proteins and has been developed for the Quality Control process of the MALDI Biotyper system. Its specific composition covers the entire mass range of proteins used by MALDI Biotyper for precise identification of microorganisms, making BTS ideal for use with this mass spectrometry system. 5 tubes, each tube sufficient for approximately 40 quality controls