Learn More
Hach Company Brush, Buret, 25 mL
Supplier:  Hach Company 68500
Encompass
Ideally shaped for quick and thorough cleaning of burets and other similarly shaped vessels. Unbleached white nylon bristles are supported by twisted galvanized steel wire. Brush portion measures 8 cm (3.1") in length. Fan style tip. 1.3cm (0.5") Dia. 76cm (30") L
Catalog No. 50-199-4426