Learn More
Gilson Company Inc BRUSH COARSE SIEVE BRASS
Supplier:  Gilson Company Inc TSA172
Encompass_Preferred
Coarse Sieve Brush, 8-1/2 in. , Curved Plastic Handle, 1-1/2 in. X 1-3/4 in. Slanted Brass Wire Bristles, Coarser Wire Cloth In Round Sieves.
Catalog No. NC9019265
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.