Learn More
Med Vet International Sterile Surgical Scrub Brush Dry Sterile, Each
Supplier:  Med Vet International BR125
Encompass_Preferred
Soft outer bristles clean thoroughly without skin irritation. Inner bristles are shorter and contoured for effective deep cleaning of fingers, nails and cuticles. Brush flexes for easier scrubbing between and around fingers. Convenient peel-open package.
Catalog No. 50-252-2433