Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Era Enterprises BRUSH FOR 5MM O.D THIN WALL

Supplier:  New Era Enterprises NE3435

Encompass_Preferred

Brush for 5mm OD thin wall tubes, 50mm of bristles, 240mm OAL

Catalog No. NC9382159


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.