Learn More
Chemglass Life Sciences Brush, Glassware Cleaning, Test Tubes
SDP
Supplier:  Chemglass Life Sciences CG114301
Encompass_Preferred
Brush, Glassware Cleaning, Test Tubes. Brush has polypropylene bristles and can be spread apart to clean tubes having an I.D. of 14mm or greater. MADE IN USA.
Catalog No. 50-131-0996