Learn More
Chemglass Life Sciences Foam Brush Set, Glassware Cleaning
SDP
Supplier:  Chemglass Life Sciences CG114101
Encompass_Preferred
Soft plush foam gently cleans interior glassware surfaces without absorbing water. Bristles never become soggy or limp. Lasts up to 20 times longer than commonly used sponges. Supplied complete with 3 brushes. Set includes 1 of each brush: 9” Overall Length, Foam Ball 2” in Diameter, 17” Overall Length, Foam Ball 2” in Diameter, 18” Overall Length, Fits Through a 3/4” Opening.
Catalog No. 50-131-0993