Learn More
Apexbio Technology LLC BS-181 HCl, 10mM (in 1mL DMSO) Cas# 1397219-81-6
SDP
Supplier:  Apexbio Technology LLC A57005.1
Encompass_Preferred
highly selective CDK7 inhibitor 1397219-81-6. MFCD18384975 IC50: 21 nm (CDK7) Normal progression through the cell cycle requires the sequential action of cyclin-dependent kinases CDK1, CDK2, CDK4, and CDK6. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8589