Learn More
Bioworld Bovine Serum Albumin (BSA), 30%, Sterile, 500 mL
Supplier:  Bioworld 402200602
Encompass_Preferred
Biotechnology Grade Bovine Serum Albumin:A general purpose purified albumin, suitable for Westerns, enzyme systems and as a protein supplementAppearance: LiquidPH Value: 6.40-7.40
Catalog No. 50-255-470