Learn More
Bioworld BSA BLOCKING BUFFER IN TBS
Supplier:  Bioworld 402200952
Encompass_Preferred
Blot blocking buffer used to block non-specific binding site for antibodies on the membrane. Specification: 3% BSA in TBS with 0.02% Sodium azide.
Catalog No. 50-255-488