Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Standard Grade, 25 g
Supplier:  Bioworld 220700081
Encompass_Preferred
BSA (Bovine Serum Albumin), Fraction VProtein with numerous biochemical applications including ELISAs, immunoblots, and immunohistochemistryHeat shock isolationSpecifications:pH (5% water): 6.5-7.5 Heavy Metals (Pb): <0.001% DNase: None detected RNase: None detected Protease: None detectedUsually ships within 24 hours. This product is sold for laboratory use only.
Catalog No. 50-253-899