Learn More
Bioworld Bovine Serum Albumin (BSA), 1 mg/mL, 25 mL
Supplier:  Bioworld 220700051
Encompass_Preferred
1mg/ml BSA (Bovine serum albumin) solution in 0.85% Sodium Chloride and 0.01% Sodium azide.
Catalog No. 50-253-894