Learn More
Cygnus Technologies BSA Sample Diluent, 100mL
Supplier:  Cygnus Technologies F031A100
Encompass
For sample dilution with Cygnus BSA, Bovine IgG, Bovine Plasma Protein, and Bovine Transferrin ELISA Kits.
Catalog No. NC9908769