Learn More
Cygnus Technologies BSA ELISA KIT
Supplier:  Cygnus Technologies F030
Encompass
This is a very sensitive assay for BSA, with a limit of detection of <250pg/mL. This kit can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Catalog No. NC9580120