Learn More
Gold Biotechnology Inc BSA Fraction V Fatty Acid Free
SDP
Supplier:  Gold Biotechnology Inc A42125
Encompass_Preferred
GoldBio s Reagent Grade Fatty Acid Free BSA is ideally suited for fatty acid sensitive cell culture systems such as CHO Vero and MDCK cells as well as for the most sensitive diagnostic and research applications
Catalog No. 50-005-62172